2-Ethoxy-4-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A644067-1G A644067-250MG
77198-402EA 205 USD
77198-402 77198-404
2-Ethoxy-4-fluorobenzoic acid 97%
2-Ethoxy-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD14582790
CAS Number: 1233541-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR