1,3-Diacetylbenzene 97%

Supplier: Ambeed
Warning

A643961-100G A643961-1G A643961-25G A643961-5G
77198-394EA 271.01 USD
77198-394 77198-396 77198-398 77198-400
1,3-Diacetylbenzene 97%
1,3-Diacetylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00008740
CAS Number: 6781-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR