6-(tert-Butyl)pyridin-3-amine 95%

Supplier: Ambeed
Warning

A643142-100MG A643142-1G A643142-250MG
77170-360EA 106.09 USD
77170-360 77170-362 77170-364
6-(tert-Butyl)pyridin-3-amine 95%
6-(tert-Butyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.22 g/mol
MDL Number: MFCD12033372
CAS Number: 39919-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR