3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4'-methoxy-[1,1'-biphenyl]-4-amine

A642226-100MG A642226-1G
77198-384EA 38.11 USD
77198-384 77198-386
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine 97%
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FNO
MW: 217.25 g/mol
MDL Number: MFCD06802177
CAS Number: 893734-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR