2-Chloro-3,4-diiodopyridine 95+%

Supplier: Ambeed
Warning

A641172-1G A641172-250MG A641172-5G
77170-352EA 86.52 USD
77170-352 77170-354 77170-356
2-Chloro-3,4-diiodopyridine 95+%
2-Chloro-3,4-diiodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClI₂N
MW: 365.33 g/mol
MDL Number: MFCD16878974
CAS Number: 153034-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR