1-benzyl-1,4-diazepane 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylhomopiperazine, 1-Benzyl-1,4-diazepane

A640488-1G A640488-250MG A640488-5G
77198-366EA 29.87 USD
77198-366 77198-368 77198-370
1-benzyl-1,4-diazepane 95%
1-benzyl-1,4-diazepane

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD00800858
CAS Number: 4410-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR