3-Hydroxy-4-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A640022-1G A640022-5G A640022-250MG
77170-350EA 67 USD
77170-350 77573-066 77893-280
3-Hydroxy-4-iodobenzonitrile 95%
3-Hydroxy-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄INO
MDL Number: MFCD12828346
CAS Number: 210962-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR