5,6-Dimethoxybenzo[b]thiophene 97%

Supplier: Ambeed
Warning

A638804-100MG A638804-1G A638804-250MG
77198-334EA 85 USD
77198-334 77198-336 77198-338
5,6-Dimethoxybenzo[b]thiophene 97%
5,6-Dimethoxybenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂S
MDL Number: MFCD00981875
CAS Number: 91715-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR