1,4-Bis[(1H-imidazol-1-yl)methyl]benzene 97%

Supplier: Ambeed
Warning

A638374-250MG A638374-5G A638374-1G
77198-330EA 12 USD
77198-330 77198-332 77747-313
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene 97%
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄
MDL Number: MFCD06598031
CAS Number: 56643-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR