5-Bromo-7H-benzo[c]carbazole 98%

Supplier: Ambeed
Warning

A636504-1G A636504-250MG A636504-25G A636504-5G
77198-314EA 37.08 USD
77198-314 77198-316 77198-318 77198-320
5-Bromo-7H-benzo[c]carbazole 98%
5-Bromo-7H-benzo[c]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₀BrN
MW: 296.17 g/mol
MDL Number: MFCD30063204
CAS Number: 131409-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR