tert-Butyl (3-bromopyridin-2-yl)carbamate 96%
Supplier: Ambeed
|
Warning
|
A636301-100MG
A636301-1G
A636301-250MG
77170-336EA
61
USD
77170-336
77170-338
77170-340
tert-Butyl (3-bromopyridin-2-yl)carbamate 96%
tert-Butyl (3-bromopyridin-2-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrN₂O₂ |
MDL Number:
MFCD11976399 CAS Number: 149489-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...