tert-Butyl-5-bromo-1H-indazole-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1H-indazole, N1-BOC protected, tert-Butyl 5-bromo-1H-indazole-1-carboxylate

A635528-10G A635528-1G A635528-5G
77170-330EA 215.27 USD
77170-330 77170-332 77170-334
tert-Butyl-5-bromo-1H-indazole-1-carboxylate 97%
tert-Butyl-5-bromo-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.16 g/mol
MDL Number: MFCD08435913
CAS Number: 651780-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR