2,4-Dimethyl-3-pyridinecarboxylic acid 95+%

Supplier: Ambeed
Warning

A635397-100MG A635397-1G A635397-250MG A635397-5G
77170-322EA 14 USD
77170-322 77170-324 77170-326 77170-328
2,4-Dimethyl-3-pyridinecarboxylic acid 95+%
2,4-Dimethyl-3-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD03265250
CAS Number: 55314-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR