3-Bromo-4-fluorobenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A634655-100MG A634655-250MG
77170-318EA 6 USD
77170-318 77170-320
3-Bromo-4-fluorobenzene-1,2-diamine 97%
3-Bromo-4-fluorobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆BrFN₂
MDL Number: MFCD18071059
CAS Number: 1257535-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR