2-Acetylamino-4-methylpyridine 98%

Supplier: Ambeed
Warning

A633949-1G A633949-250MG A633949-5G
77170-312EA 15 USD
77170-312 77170-314 77170-316
2-Acetylamino-4-methylpyridine 98%
2-Acetylamino-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD00234432
CAS Number: 5327-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR