4-(1-Amino-2,2,2-trifluoroethyl)phenol 96%

Supplier: Ambeed
Warning

A633621-100MG A633621-1G A633621-250MG
77170-306EA 105.06 USD
77170-306 77170-308 77170-310
4-(1-Amino-2,2,2-trifluoroethyl)phenol 96%
4-(1-Amino-2,2,2-trifluoroethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD09832191
CAS Number: 294175-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR