(2-Iodoethyl)cyclopropane 97% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with Copper chip

A633187-1G A633187-250MG A633187-5G
77170-300EA 145.23 USD
77170-300 77170-302 77170-304
(2-Iodoethyl)cyclopropane 97% stabilized
(2-Iodoethyl)cyclopropane

Precaución: for Research Use Only

Formula: C₅H₉I
MW: 196.02 g/mol
MDL Number: MFCD08669937
CAS Number: 335449-19-9
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR