5-Bromo-1,3-difluoro-2-propoxybenzene 97%

Supplier: Ambeed
Warning

A632573-100MG A632573-1G A632573-250MG
77198-290EA 39 USD
77198-290 77198-292 77198-294
5-Bromo-1,3-difluoro-2-propoxybenzene 97%
5-Bromo-1,3-difluoro-2-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.06 g/mol
MDL Number: MFCD18917885
CAS Number: 1309933-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR