2-(1H-Imidazol-5-yl)ethanol 95%

Supplier: Ambeed
Warning

A632520-100MG A632520-1G A632520-5G A632520-250MG
77170-294EA 10.3 USD
77170-294 77170-296 77170-298 77684-517
2-(1H-Imidazol-5-yl)ethanol 95%
2-(1H-Imidazol-5-yl)ethanol

Precaución: For research use only.

Formula: C₅H₈N₂O
MW: 112.1300 g/mol
MDL Number: MFCD00195671
CAS Number: 872-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR