Methyl 5-bromo-4-fluoro-2-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
A632275-100MG
A632275-1G
A632275-250MG
77198-284EA
7
USD
77198-284
77198-286
77198-288
Methyl 5-bromo-4-fluoro-2-hydroxybenzoate 98%
Methyl 5-bromo-4-fluoro-2-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₃ |
MDL Number:
MFCD27939807 CAS Number: 4133-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...