2,6-Dichloro-4-(difluoromethyl)nicotinonitrile 95%

Supplier: Ambeed
Warning

A632047-100MG A632047-1G A632047-250MG
77170-288EA 64.89 USD
77170-288 77170-290 77170-292
2,6-Dichloro-4-(difluoromethyl)nicotinonitrile 95%
2,6-Dichloro-4-(difluoromethyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₂N₂
MW: 223.00 g/mol
MDL Number: MFCD25486811
CAS Number: 960293-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR