PRIMA-1MET 97%

Supplier: Ambeed
Warning

Synonyms: Eprenetapopt, PRIMA-1Met, 2-(Hydroxymethyl)-2-(methoxymethyl)quinuclidin-3-one, APR-246, 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one

A631504-100MG A631504-10MG A631504-1MG A631504-25MG A631504-50MG A631504-5MG
77198-272EA 365.65 USD
77198-272 77198-274 77198-276 77198-278 77198-280 77198-282
PRIMA-1MET 97%
PRIMA-1MET

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD16294187
CAS Number: 5291-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR