2,5-Dichloro-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloropyridine-3-boronic acid

A630324-1G A630324-250MG A630324-5G
77170-278EA 129.78 USD
77170-278 77170-280 77170-282
2,5-Dichloro-3-pyridineboronic acid 98%
2,5-Dichloro-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
MDL Number: MFCD06659486
CAS Number: 536693-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR