4-Chloro-2,6-dibromoanisole 97%

Supplier: Ambeed
Warning

A627554-100MG A627554-1G A627554-250MG
77198-250EA 34 USD
77198-250 77198-252 77198-254
4-Chloro-2,6-dibromoanisole 97%
4-Chloro-2,6-dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₅Br₂ClO
MDL Number: MFCD00068569
CAS Number: 174913-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR