4-Bromo-5-methylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A627422-100MG A627422-250MG A627422-1G A627422-5G
77170-262EA 32 USD
77170-262 77170-264 77893-261 77893-262
4-Bromo-5-methylpyridin-2(1H)-one 95%
4-Bromo-5-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MDL Number: MFCD16610029
CAS Number: 1227578-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR