Dicyclohexyl carbonate 95%

Supplier: Ambeed
Warning

A625569-5G A625569-25G A625569-1G
77170-254EA 66 USD
77170-254 77893-256 77893-255
Dicyclohexyl carbonate 95%
Dicyclohexyl carbonate

Precaución: for Research Use Only

Formula: C₁₃H₂₂O₃
MDL Number: MFCD28718007
CAS Number: 4427-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR