2-(((2-Aminoethyl)(methyl)amino)methyl)benzonitrile 97%
Supplier: Ambeed
|
Warning
|
A623535-100MG
A623535-1G
A623535-250MG
A623535-50MG
77198-230EA
34
USD
77198-230
77198-232
77198-234
77198-236
2-(((2-Aminoethyl)(methyl)amino)methyl)benzonitrile 97%
2-(((2-Aminoethyl)(methyl)amino)methyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₁H₁₅N₃ |
MDL Number:
MFCD16846301 CAS Number: 1249299-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...