1,12-Diaminododecane 98%

Supplier: Ambeed
Danger

A622495-5G A622495-25G
77198-224EA 8 USD
77198-224 77573-020
1,12-Diaminododecane 98%
1,12-Diaminododecane

Precaución: for Research Use Only

Formula: C₁₂H₂₈N₂
MDL Number: MFCD00008155
CAS Number: 2783-17-7
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR