(+)-N,N,N',N'-Tetramethyl-L-tartardiamide 98%

Supplier: Ambeed
Warning

A621744-100G A621744-10G A621744-1G A621744-25G A621744-5G
77198-214EA 356.38 USD
77198-214 77198-216 77198-218 77198-220 77198-222
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide 98%
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄
MDL Number: MFCD00025672
CAS Number: 26549-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR