1-(4-Methylphenyl)-2-thiourea 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methylphenylthiourea, N-(p-Tolyl)thiourea, N-(4-Methylphenyl)thiourea, p-Tolylthiourea, 1-(p-Tolyl)thiourea, 1-(4-Tolyl)thiourea

A621741-100G A621741-10G A621741-1G A621741-25G A621741-5G
77198-204EA 194.67 USD
77198-204 77198-206 77198-208 77198-210 77198-212
1-(4-Methylphenyl)-2-thiourea 98%
1-(4-Methylphenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₄(CH₃))NC(S)NH₂
MW: 166.25 g/mol
MDL Number: MFCD00041190
CAS Number: 622-52-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR