Methyl 4-bromo-2-chloro-5-fluorobenzoate 95%

Supplier: Ambeed
Warning

A621344-1G A621344-250MG A621344-5G
77170-234EA 47 USD
77170-234 77170-236 77170-238
Methyl 4-bromo-2-chloro-5-fluorobenzoate 95%
Methyl 4-bromo-2-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
MDL Number: MFCD12913461
CAS Number: 908248-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR