(3-(Cyclopropylcarbamoyl)-2-fluorophenyl)boronic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid

A620729-1G A620729-250MG
77198-200EA 81 USD
77198-200 77198-202
(3-(Cyclopropylcarbamoyl)-2-fluorophenyl)boronic acid 97%
(3-(Cyclopropylcarbamoyl)-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BFNO₃
MW: 223.00 g/mol
MDL Number: MFCD22682843
CAS Number: 2096353-44-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR