4-Bromo-3,5-dimethylbenzoic acid 98%

Supplier: Ambeed
Warning

A620510-5G A620510-1G A620510-250MG A620510-25G
77170-230EA 82 USD
77170-230 77893-247 77893-248 77893-249
4-Bromo-3,5-dimethylbenzoic acid 98%
4-Bromo-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD16250111
CAS Number: 7697-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR