(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

A619511-100MG A619511-1G A619511-250MG
77170-214EA 52.53 USD
77170-214 77170-216 77170-218
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-2-methylpropanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
MW: 341.35 g/mol
MDL Number: MFCD02682594
CAS Number: 207117-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR