7-Bromo-2-methylbenzo[d]thiazole 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A619365-250MG A619365-5G
77170-212EA 84 USD
77170-212 77684-498
7-Bromo-2-methylbenzo[d]thiazole 95%, Ambeed.Inc
7-Bromo-2-methylbenzo[d]thiazole

Precaución: For research use only.

Formula: C₈H₆BrNS
MDL Number: MFCD11217386
CAS Number: 110704-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR