10-Aminodecan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 10-Amino-1-decanol

A618981-100MG A618981-1G A618981-250MG A618981-5G
77198-192EA 42.23 USD
77198-192 77198-194 77198-196 77198-198
10-Aminodecan-1-ol 98%
10-Aminodecan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₂₃NO
MW: 173.2957 g/mol
MDL Number: MFCD02094313
CAS Number: 23160-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR