5-Cyano-2-fluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-2-fluorophenylboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A616906-1G A616906-250MG A616906-5G
77198-174EA 81.37 USD
77198-174 77198-176 77198-178
5-Cyano-2-fluorobenzeneboronic acid pinacol ester 98%
5-Cyano-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
MDL Number: MFCD07437899
CAS Number: 863868-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR