5-Bromo-2-fluoro-1-methoxy-3-methylbenzene 97%

Supplier: Ambeed
Warning

A616668-1G A616668-250MG
77198-170EA 33 USD
77198-170 77198-172
5-Bromo-2-fluoro-1-methoxy-3-methylbenzene 97%
5-Bromo-2-fluoro-1-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MDL Number: MFCD20526392
CAS Number: 1351668-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR