1,3,5-Tribenzoylbenzene 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzene-1,3,5-triyltris(phenylmethanone)
A615892-1G
A615892-5G
77198-166EA
56
USD
77198-166
77198-168
1,3,5-Tribenzoylbenzene 95%
1,3,5-Tribenzoylbenzene
Precaución: for Research Use Only
Formula:
C₂₇H₁₈O₃ |
MDL Number:
MFCD00016577 CAS Number: 25871-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...