1-Acetyl-5-bromoindole 97%

Supplier: Ambeed
Warning

A615320-100MG A615320-1G A615320-5G
77198-160EA 16.48 USD
77198-160 77198-162 77198-164
1-Acetyl-5-bromoindole 97%
1-Acetyl-5-bromoindole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD00238530
CAS Number: 61995-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR