1-[(Dibenzylamino)methyl]cyclopropanol 98%

Supplier: Ambeed
Warning

A615061-100MG A615061-1G A615061-250MG A615061-5G
77198-152EA 15.45 USD
77198-152 77198-154 77198-156 77198-158
1-[(Dibenzylamino)methyl]cyclopropanol 98%
1-[(Dibenzylamino)methyl]cyclopropanol

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO
MW: 267.36 g/mol
MDL Number: MFCD07367843
CAS Number: 428855-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR