Methyl 3-(2,5-dichlorophenyl)-3-oxopropanoate 97%

Supplier: Ambeed
Warning

A613309-1G A613309-250MG
77198-136EA 41.2 USD
77198-136 77198-138
Methyl 3-(2,5-dichlorophenyl)-3-oxopropanoate 97%
Methyl 3-(2,5-dichlorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.07 g/mol
MDL Number: MFCD07778433
CAS Number: 56719-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR