2-Cyanoisonicotinohydrazide 95%

Supplier: Ambeed
Warning

A611015-250MG A611015-5G
77198-126EA 15 USD
77198-126 77198-128
2-Cyanoisonicotinohydrazide 95%
2-Cyanoisonicotinohydrazide

Precaución: for Research Use Only

Formula: C₇H₆N₄O
MDL Number: MFCD23132274
CAS Number: 135048-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR