Ethyl 3-bromo-5-chlorobenzoate 97%

Supplier: Ambeed
Warning

A609953-100MG A609953-1G A609953-250MG A609953-5G
77198-106EA 25.75 USD
77198-106 77198-108 77198-110 77198-112
Ethyl 3-bromo-5-chlorobenzoate 97%
Ethyl 3-bromo-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
MDL Number: MFCD10566220
CAS Number: 1095274-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR