6-tert-Butyl-nicotinonitrile 95+%

Supplier: Ambeed
Danger

A609224-100MG A609224-1G A609224-250MG
77170-184EA 15 USD
77170-184 77170-186 77170-188
6-tert-Butyl-nicotinonitrile 95+%
6-tert-Butyl-nicotinonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MDL Number: MFCD11845758
CAS Number: 56029-45-9
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR