N-Methoxy-N-methyl-1H-pyrrole-2-carboxamide 98%

Supplier: Ambeed
Warning

A608964-100MG A608964-1G A608964-250MG
77198-092EA 15.45 USD
77198-092 77198-094 77198-096
N-Methoxy-N-methyl-1H-pyrrole-2-carboxamide 98%
N-Methoxy-N-methyl-1H-pyrrole-2-carboxamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD12406910
CAS Number: 368211-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR