2,4-Difluorobenzimidamide hydrochloride 98%

Supplier: Ambeed
Warning

A608627-1G A608627-5G
77198-088EA 123.6 USD
77198-088 77198-090
2,4-Difluorobenzimidamide hydrochloride 98%
2,4-Difluorobenzimidamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇ClF₂N₂
MW: 192.59 g/mol
MDL Number: MFCD04114445
CAS Number: 1170536-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR