1-Benzyl-3-methylimidazolium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium chloride

A608563-100G A608563-25G A608563-5G
77198-082EA 125.66 USD
77198-082 77198-084 77198-086
1-Benzyl-3-methylimidazolium chloride 98%
1-Benzyl-3-methylimidazolium chloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂
MW: 208.69 g/mol
MDL Number: MFCD07784451
CAS Number: 36443-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR