1H-Pyrrole-2,3-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A608456-100MG A608456-1G A608456-250MG
77198-076EA 19 USD
77198-076 77198-078 77198-080
1H-Pyrrole-2,3-dicarboxylic acid 97%
1H-Pyrrole-2,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₄
MDL Number: MFCD09991931
CAS Number: 1125-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR