5-Fluoroquinazoline 97%

Supplier: Ambeed
Warning

A607897-1G A607897-250MG
77198-068EA 325.48 USD
77198-068 77198-070
5-Fluoroquinazoline 97%
5-Fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₅FN₂
MW: 148.14 g/mol
MDL Number: MFCD18448849
CAS Number: 16499-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR